Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000257.3 | NP_000248.2 | 39, 40 | 5929 | ||
XM_017021340.1 | XP_016876829.1 | 38, 39 | 5864 | |||
GenBank mRNA | AB209708.1 | - | 28, 29 | 4716 | ||
AJ710171.1 | - | 3, 4 | 321 | |||
AY518538.1 | - | 20, 21 | 3531 | |||
BC112171.1 | - | 38, 39 | 5853 | 8327639 | ||
BC112173.1 | - | 38, 39 | 5853 | 8327641 | ||
DQ248310.1 | - | 17, 18 | 2901 | |||
EF560725.1 | - | 37, 38 | 5796 | |||
EU747717.1 | - | 39, 40 | 5904 | |||
M17712.1 | - | 9, 10 | 1624 | |||
M21665.1 | - | 9, 10 | 1565 | |||
M58018.1 | - | 39, 40 | 5884 | |||
X03741.1 | - | 11, 12 | 1898 | |||
X05631.1 | - | 2, 3 | 239 | |||
X06976.1 | - | 9, 10 | 1624 | |||
X51591.1 | - | 19, 20 | 3446 |