Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000432.3 | NP_000423.2 | 7 | 801 | ||
GenBank mRNA | BC015821.1 | - | 7 | 783 | 4245364 | |
BC031008.1 | - | 7 | 780 | 4297454 | ||
X66141.1 | - | 7 | 760 |