Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000433.3 | NP_000424.2 | 3 | 595 | ||
NM_001127651.2 | NP_001121123.1 | 4 | 432 | |||
NM_001190789.1 | NP_001177718.1 | 3 | 595 | |||
NM_001190794.1 | NP_001177723.1 | 3 | 595 | |||
XM_005245207.1 | XP_005245264.1 | 3 | 524 | |||
XM_011509580.1 | XP_011507882.1 | 4 | 433 | |||
XM_011509581.1 | XP_011507883.1 | 4 | 420 | |||
GenBank mRNA | AB209647.1 | - | 3 | 475 | ||
AF527950.1 | - | 3 | 342 | |||
AK296672.1 | - | 3 | 451 | |||
AK298713.1 | - | 3 | 504 | |||
AK312666.1 | - | 3 | 505 | |||
BC001606.1 | - | 3 | 572 | 3542841 | ||
BP193903.1 | - | 4 | 279 | |||
BT007439.1 | - | 3 | 320 | |||
DA420141.1 | - | 4 | 423 | |||
DA817767.1 | - | 4 | 433 | |||
KU178216.1 | - | 3 | 320 | |||
M32011.1 | - | 4 | 387 |