Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KITLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000899.4 | NP_000890.1 | 10 | 1332 | ||
NM_003994.5 | NP_003985.2 | 9 | 1248 | |||
GenBank mRNA | AF400436.1 | - | 10 | 1321 | ||
BC126166.1 | - | 10 | 1287 | 8991882 | ||
BC143899.1 | - | 9 | 1203 | 9052417 | ||
M59964.1 | - | 10 | 1321 |