Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IPO5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002271.4 | NP_002262.3 | 5 | 384 | ||
XM_005254052.3 | XP_005254109.1 | 4 | 294 | |||
XM_005254053.3 | XP_005254110.1 | 4 | 311 | |||
XM_011521087.2 | XP_011519389.1 | 6 | 475 | |||
XM_011521088.2 | XP_011519390.1 | 6 | 541 | |||
XM_011521089.2 | XP_011519391.1 | 7 | 598 | |||
XM_011521090.2 | XP_011519392.1 | 5 | 434 | |||
XM_017020562.1 | XP_016876051.1 | 5 | 415 | |||
GenBank mRNA | AK225286.1 | - | 2 | 169 | ||
AK302812.1 | - | 2 | 156 | |||
AK303494.1 | - | 2 | 172 | |||
BC001497.2 | - | 2 | 152 | 2823581 | ||
BC019309.1 | - | 2 | 152 | |||
BC045640.1 | - | 5 | 365 | 5298410 | ||
BX648222.1 | - | 2 | 214 | |||
U72761.1 | - | 2 | 150 | |||
Y08890.1 | - | 5 | 386 |