Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IPO5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002271.4 | NP_002262.3 | 8 | 765 | ||
XM_005254052.3 | XP_005254109.1 | 7 | 675 | |||
XM_005254053.3 | XP_005254110.1 | 7 | 692 | |||
XM_011521087.2 | XP_011519389.1 | 9 | 856 | |||
XM_011521088.2 | XP_011519390.1 | 9 | 922 | |||
XM_011521089.2 | XP_011519391.1 | 10 | 979 | |||
XM_011521090.2 | XP_011519392.1 | 8 | 815 | |||
XM_017020562.1 | XP_016876051.1 | 8 | 796 | |||
GenBank mRNA | AK225286.1 | - | 5 | 550 | ||
AK302812.1 | - | 5 | 537 | |||
BC001497.2 | - | 5 | 533 | 2823581 | ||
BC019309.1 | - | 5 | 533 | |||
BC045640.1 | - | 8 | 746 | 5298410 | ||
BX648222.1 | - | 5 | 595 | |||
U72761.1 | - | 5 | 531 | |||
Y08890.1 | - | 8 | 767 |