Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002272.3 | NP_002263.3 | 2 | 660 | ||
GenBank mRNA | AK056254.1 | - | 2 | 1137 | ||
AK098110.1 | - | 2 | 688 | |||
AK296582.1 | - | 3 | 627 | |||
AK299394.1 | - | 3 | 603 | |||
AK299400.1 | - | 2 | 660 | |||
AK299451.1 | - | 3 | 522 | |||
AK307941.1 | - | 2 | 407 | |||
BC003630.2 | - | 1 | 16 | |||
BC042174.1 | - | 665 | 5453644 | |||
X07695.1 | - | 2 | 266 |