Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300810.1 | NP_001287739.1 | 7 | 1559 | ||
NM_002283.3 | NP_002274.1 | 9 | 2113 | |||
GenBank mRNA | X99140.1 | - | 9 | 2059 |