Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002373.5 | NP_002364.5 | 4 | 1407 | ||
XM_017022189.1 | XP_016877678.1 | 5 | 1697 | |||
GenBank mRNA | AF095910.1 | - | 1 | 67 | ||
AF200415.2 | - | 2 | 1139 | |||
AK291052.1 | - | 4 | 1379 | |||
BC055312.1 | - | 4 | 1376 | |||
BC094702.1 | - | 4 | 1366 | |||
U38292.1 | - | 1 | 940 | |||
U80458.1 | - | 1 | 709 |