Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002427.3 | NP_002418.1 | 10 | 1395 | ||
GenBank mRNA | AK222748.1 | - | 10 | 1395 | ||
AK292211.1 | - | 10 | 1395 | |||
BC067522.1 | - | 10 | 1374 | 7002184 | ||
BC067523.1 | - | 10 | 1374 | 7002185 | ||
BC074807.2 | - | 10 | 1385 | 30915315 | ||
BC074808.2 | - | 10 | 1385 | 30915529 | ||
X75308.1 | - | 10 | 1370 | |||
X81334.1 | - | 10 | 1393 |