Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEACAM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002483.6 | NP_002474.4 | 6 | 2090 | ||
XM_011526990.1 | XP_011525292.1 | 5 | 2072 | |||
GenBank mRNA | BC005008.1 | - | 6 | 2011 | 3640231 | |
M18216.1 | - | 6 | 2015 | |||
M18728.1 | - | 6 | 2021 |