Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPTX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002522.3 | NP_002513.2 | 5 | 1981 | ||
GenBank mRNA | AK127335.1 | - | 5 | 1981 | ||
U61849.1 | - | 5 | 1951 |