Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPTX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002522.3 | NP_002513.2 | 5 | 5010 | ||
GenBank mRNA | AK124701.1 | - | 4 | 3844 | ||
AK127335.1 | - | 5 | 5010 | |||
BX537550.1 | - | 2 | 5752 | |||
CK005561.1 | - | 1 | 110 | |||
U61849.1 | - | 5 | 4950 |