Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPTX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002523.2 | NP_002514.1 | 3 | 849 | ||
GenBank mRNA | AK311250.1 | - | 2 | 540 | ||
BC009924.2 | - | 3 | 841 | 3508049 | ||
BC034781.1 | - | 3 | 837 | 5169124 | ||
BC035339.1 | - | 3 | 842 | 5198692 | ||
BC048275.1 | - | 3 | 807 | 5165672 | ||
U26662.1 | - | 3 | 565 |