Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NTRK3 SIRNA Assays ›
Gene Symbol:
NTRK3
Gene Name:
neurotrophic receptor tyrosine kinase 3
Gene Aliases:
GP145-TrkC, TRKC, gp145(trkC)
Chromosome Location:
Chr.15: 87859749 - 88256796 on Build GRCh38
UniGene ID:
Hs.185701
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
143655
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NTRK3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001007156.2
NP_001007157.1
4
614
NM_001012338.2
NP_001012338.1
4
614
NM_001243101.1
NP_001230030.1
4
614
NM_001320134.1
NP_001307063.1
2
324
NM_001320135.1
NP_001307064.1
2
215
NM_002530.3
NP_002521.2
4
614
XM_006720543.3
XP_006720606.1
2
923
XM_006720544.3
XP_006720607.1
2
923
XM_006720545.3
XP_006720608.1
2
923
XM_006720548.3
XP_006720611.1
2
923
XM_006720549.3
XP_006720612.1
2
923
XM_006720550.3
XP_006720613.1
2
923
XM_011521637.2
XP_011519939.1
2
923
XM_011521638.2
XP_011519940.1
2
923
XM_017022240.1
XP_016877729.1
3
794
XM_017022241.1
XP_016877730.1
2
279
XM_017022242.1
XP_016877731.1
2
923
XM_017022243.1
XP_016877732.1
2
275
XM_017022244.1
XP_016877733.1
2
177
XM_017022245.1
XP_016877734.1
3
232
XM_017022246.1
XP_016877735.1
2
333
XM_017022247.1
XP_016877736.1
2
923
XM_017022248.1
XP_016877737.1
2
923
XM_017022249.1
XP_016877738.1
2
923
XM_017022250.1
XP_016877739.1
2
923
XM_017022253.1
XP_016877742.1
2
923
XM_017022254.1
XP_016877743.1
2
923
GenBank mRNA
AK297160.1
-
2
332
AK302504.1
-
2
174
AK309695.1
-
2
334
AK313646.1
-
2
333
BC013693.1
-
2
313
4181574
BT007291.1
-
2
308
HY003065.1
-
2
215
KJ534904.1
-
2
308
KJ534905.1
-
2
308
KJ534906.1
-
2
308
KJ535052.1
-
2
308
S76475.1
-
2
323
S76476.1
-
2
323
U05012.1
-
3
463
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
neuron migration
negative regulation of protein phosphorylation
positive regulation of protein phosphorylation
transmembrane receptor protein tyrosine kinase signaling pathway
heart development
circadian rhythm
positive regulation of cell proliferation
positive regulation of gene expression
peptidyl-tyrosine phosphorylation
modulation by virus of host transcription
positive regulation of cell migration
activation of protein kinase B activity
positive regulation of peptidyl-serine phosphorylation
neurotrophin signaling pathway
mechanoreceptor differentiation
positive regulation of apoptotic process
response to ethanol
protein autophosphorylation
ephrin receptor signaling pathway
neuron fate specification
response to axon injury
positive regulation of axon extension involved in regeneration
negative regulation of astrocyte differentiation
positive regulation of positive chemotaxis
response to corticosterone
positive regulation of synapse assembly
negative regulation of cell death
lens fiber cell differentiation
cellular response to retinoic acid
cochlea development
activation of GTPase activity
positive regulation of actin cytoskeleton reorganization
p53 binding
GPI-linked ephrin receptor activity
neurotrophin receptor activity
protein binding
ATP binding
neurotrophin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0