Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDGFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002608.3 | NP_002599.1 | 7 | 2830 | ||
NM_033016.3 | NP_148937.1 | 7 | 1730 | |||
GenBank mRNA | AK022920.1 | - | 1 | 934 | ||
BC029822.1 | - | 7 | 2050 | 5174750 | ||
M12783.1 | - | 7 | 2828 | |||
X02744.1 | - | 7 | 1856 | |||
X03702.1 | - | 6 | 1665 |