Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PI4KB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198773.1 | NP_001185702.1 | 4 | 1679 | ||
NM_001198774.1 | NP_001185703.1 | 2 | 1362 | |||
NM_002651.2 | NP_002642.1 | 3 | 1506 | |||
XM_005245264.3 | XP_005245321.1 | 3 | 1506 | |||
XM_011509634.1 | XP_011507936.1 | 3 | 1506 | |||
XM_011509635.1 | XP_011507937.1 | 4 | 1679 | |||
XM_011509636.2 | XP_011507938.1 | 2 | 1334 | |||
XM_011509637.2 | XP_011507939.1 | 3 | 1507 | |||
XM_011509638.1 | XP_011507940.1 | 3 | 1506 | |||
XM_017001486.1 | XP_016856975.1 | 4 | 1679 | |||
XM_017001487.1 | XP_016856976.1 | 2 | 1334 | |||
XM_017001488.1 | XP_016856977.1 | 3 | 1507 | |||
XM_017001489.1 | XP_016856978.1 | 4 | 1679 | |||
XM_017001490.1 | XP_016856979.1 | 2 | 1334 | |||
XM_017001491.1 | XP_016856980.1 | 3 | 1507 | |||
XM_017001492.1 | XP_016856981.1 | 3 | 1507 | |||
GenBank mRNA | AB005910.1 | - | 2 | 994 | ||
AJ011121.1 | - | 2 | 979 | |||
BC000029.2 | - | 2 | 1042 | 3508885 | ||
BC040300.1 | - | 2 | 1362 | 4821417 | ||
U81802.1 | - | 3 | 1178 |