Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKNOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286258.1 | NP_001273187.1 | 9 | 1064 | ||
NM_001320694.1 | NP_001307623.1 | 10 | 1233 | |||
NM_004571.4 | NP_004562.2 | 10 | 1236 | |||
GenBank mRNA | AK222892.1 | - | 9 | 1102 | ||
AK294794.1 | - | 9 | 933 | |||
AY142115.1 | - | 10 | 1213 | |||
AY196965.1 | - | 10 | 1214 | |||
BC007746.2 | - | 9 | 1043 | 4099156 | ||
U68727.1 | - | 9 | 1107 | |||
Y13613.1 | - | 10 | 1149 |