Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NRCAM SIRNA Assays ›
Gene Symbol:
NRCAM
Gene Name:
neuronal cell adhesion molecule
Gene Aliases:
-
Chromosome Location:
Chr.7: 108147626 - 108456442 on Build GRCh38
UniGene ID:
Hs.21422
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
143953
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NRCAM
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001037132.2
NP_001032209.1
21, 22
2752
NM_001193582.1
NP_001180511.1
24, 25
3132
NM_001193583.1
NP_001180512.1
23, 24
3075
NM_001193584.1
NP_001180513.1
23, 24
3075
NM_005010.4
NP_005001.3
22, 23
3084
XM_005250373.2
XP_005250430.1
24, 25
3132
XM_005250383.2
XP_005250440.1
24, 25
3132
XM_005250385.2
XP_005250442.1
24, 25
3132
XM_006716003.1
XP_006716066.1
24, 25
3132
XM_006716007.1
XP_006716070.1
24, 25
3132
XM_006716012.1
XP_006716075.1
24, 25
3132
XM_006716014.1
XP_006716077.1
24, 25
3132
XM_011516253.1
XP_011514555.1
24, 25
3132
XM_011516255.1
XP_011514557.1
23, 24
2887
XM_011516256.1
XP_011514558.1
24, 25
3132
XM_011516257.1
XP_011514559.1
23, 24
3114
XM_011516258.1
XP_011514560.1
22, 23
2869
XM_011516259.1
XP_011514561.1
23, 24
3102
XM_011516261.1
XP_011514563.1
22, 23
3084
XM_011516262.1
XP_011514564.1
23, 24
3075
XM_011516265.2
XP_011514567.1
24, 25
3132
XM_011516266.2
XP_011514568.1
24, 25
3132
XM_011516267.1
XP_011514569.1
24, 25
3132
XM_011516268.2
XP_011514570.1
22, 23
2947
XM_011516269.1
XP_011514571.1
24, 25
3132
XM_011516270.1
XP_011514572.1
24, 25
3132
XM_011516271.1
XP_011514573.1
22, 23
3084
XM_017012236.1
XP_016867725.1
23, 24
3102
XM_017012237.1
XP_016867726.1
24, 25
3132
XM_017012238.1
XP_016867727.1
23, 24
3075
XM_017012239.1
XP_016867728.1
22, 23
3057
XM_017012240.1
XP_016867729.1
23, 24
3075
XM_017012241.1
XP_016867730.1
22, 23
2799
XM_017012242.1
XP_016867731.1
22, 23
2799
XM_017012243.1
XP_016867732.1
24, 25
3132
XM_017012244.1
XP_016867733.1
24, 25
3132
XM_017012245.1
XP_016867734.1
24, 25
3132
XM_017012246.1
XP_016867735.1
23, 24
3102
XM_017012247.1
XP_016867736.1
23, 24
3075
XM_017012248.1
XP_016867737.1
23, 24
3102
XM_017012249.1
XP_016867738.1
23, 24
3075
XM_017012250.1
XP_016867739.1
22, 23
2799
XM_017012251.1
XP_016867740.1
23, 24
3102
XM_017012252.1
XP_016867741.1
23, 24
3102
XM_017012253.1
XP_016867742.1
21, 22
2801
XM_017012254.1
XP_016867743.1
23, 24
3102
XM_017012255.1
XP_016867744.1
21, 22
2842
XM_017012256.1
XP_016867745.1
21, 22
2801
XM_017012257.1
XP_016867746.1
22, 23
2830
XM_017012258.1
XP_016867747.1
22, 23
2826
XM_017012259.1
XP_016867748.1
20, 21
2771
GenBank mRNA
AB002341.1
-
23, 24
3003
AJ001057.1
-
21, 22
2631
AK299870.1
-
24, 25
3120
BC114570.1
-
23, 24
2999
40083055
BC115736.1
-
23, 24
2999
40083062
BX538010.1
-
22, 23
3029
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
neuron migration
axonogenesis
axonal fasciculation
synapse assembly
central nervous system development
protein localization
single organismal cell-cell adhesion
neuronal action potential propagation
regulation of axon extension
retinal ganglion cell axon guidance
heterotypic cell-cell adhesion
clustering of voltage-gated sodium channels
positive regulation of neuron differentiation
protein binding
ankyrin binding
protein binding involved in heterotypic cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0