Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005560.4 | NP_005551.3 | 62 | 8484 | ||
XM_006723796.3 | XP_006723859.1 | 62 | 8487 | |||
XM_011528818.2 | XP_011527120.1 | 61 | 8349 | |||
XM_011528819.2 | XP_011527121.1 | 60 | 8214 | |||
GenBank mRNA | AB011105.1 | - | 17 | 2269 | ||
AF443072.1 | - | 62 | 8484 | |||
Z95636.1 | - | 2 | 191 |