Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPRIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005898.4 | NP_005889.3 | 3, 4 | 473 | ||
NM_203364.2 | NP_976240.1 | 3, 4 | 473 | |||
XM_017017749.1 | XP_016873238.1 | 2, 3 | 444 | |||
GenBank mRNA | AK127972.1 | - | 2, 3 | 273 | ||
AK299720.1 | - | 2, 3 | 348 | |||
BC001731.1 | - | 3, 4 | 473 | 3355481 | ||
Z48042.1 | - | 2, 3 | 326 |