Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MDH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199111.1 | NP_001186040.1 | 9 | 1126 | ||
NM_001199112.1 | NP_001186041.1 | 8 | 1029 | |||
NM_001316374.1 | NP_001303303.1 | 9 | 1330 | |||
NM_005917.3 | NP_005908.1 | 9 | 1330 | |||
GenBank mRNA | AK295931.1 | - | 9 | 1044 | ||
AK300719.1 | - | 8 | 855 | |||
AK312331.1 | - | 9 | 971 | |||
BC001484.2 | - | 9 | 951 | 3505345 | ||
CR457405.1 | - | 8 | 895 | |||
D55654.1 | - | 9 | 950 | |||
EU794635.1 | - | 9 | 951 | |||
U20352.1 | - | 8 | 895 |