Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIPF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198828.1 | NP_001185757.1 | 8 | 876 | ||
NM_001198829.1 | NP_001185758.1 | 9 | 1056 | |||
NM_001198830.1 | NP_001185759.1 | 9 | 957 | |||
NM_004190.3 | NP_004181.1 | 8 | 975 | |||
XM_011540311.1 | XP_011538613.1 | 8 | 917 | |||
GenBank mRNA | A01046.1 | - | 8 | 907 | ||
AK301310.1 | - | 8 | 825 | |||
AK301320.1 | - | 9 | 1004 | |||
AK308224.1 | - | 8 | 923 | |||
AK312940.1 | - | 8 | 922 | |||
AL833751.1 | - | 9 | 888 | |||
BC112272.1 | - | 7 | 894 | 8327740 | ||
BC113711.1 | - | 7 | 883 | 8322763 | ||
X05997.1 | - | 8 | 905 |