Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297418.1 | NP_001284347.1 | 9 | 1083 | ||
NM_005968.4 | NP_005959.2 | 10 | 1045 | |||
NM_031203.3 | NP_112480.2 | 11 | 928 | |||
XM_005272478.2 | XP_005272535.1 | 10 | 1389 | |||
XM_005272479.2 | XP_005272536.1 | 10 | 1389 | |||
XM_005272480.2 | XP_005272537.1 | 11 | 1272 | |||
XM_005272481.1 | XP_005272538.1 | 9 | 991 | |||
XM_005272483.2 | XP_005272540.1 | 10 | 856 | |||
XM_017026823.1 | XP_016882312.1 | 10 | 1389 | |||
XM_017026824.1 | XP_016882313.1 | 11 | 1272 | |||
XM_017026825.1 | XP_016882314.1 | 11 | 1272 | |||
XM_017026826.1 | XP_016882315.1 | 9 | 936 | |||
XM_017026827.1 | XP_016882316.1 | 9 | 991 | |||
XM_017026828.1 | XP_016882317.1 | 10 | 823 | |||
XM_017026829.1 | XP_016882318.1 | 10 | 874 | |||
XM_017026830.1 | XP_016882319.1 | 10 | 874 | |||
XM_017026831.1 | XP_016882320.1 | 10 | 843 | |||
XM_017026832.1 | XP_016882321.1 | 10 | 843 | |||
GenBank mRNA | AB209733.1 | - | 9 | 911 | ||
AF061832.1 | - | 11 | 1078 | |||
AK309659.1 | - | 994 | ||||
BC000138.2 | - | 10 | 1006 | 2900532 | ||
BC008895.2 | - | 1 | 51 | |||
BC019580.1 | - | 10 | 974 | 4938259 | ||
L03532.1 | - | 10 | 973 | |||
L32611.1 | - | 11 | 857 | |||
X72018.1 | - | 6 | 611 |