Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297418.1 | NP_001284347.1 | 10, 11 | 1193 | ||
NM_005968.4 | NP_005959.2 | 12, 13 | 1200 | |||
NM_031203.3 | NP_112480.2 | 13, 14 | 1083 | |||
XM_005272478.2 | XP_005272535.1 | 11, 12 | 1499 | |||
XM_005272480.2 | XP_005272537.1 | 12, 13 | 1382 | |||
XM_005272481.1 | XP_005272538.1 | 11, 12 | 1146 | |||
XM_005272483.2 | XP_005272540.1 | 12, 13 | 1011 | |||
XM_017026828.1 | XP_016882317.1 | 11, 12 | 933 | |||
XM_017026831.1 | XP_016882320.1 | 12, 13 | 998 | |||
XM_017026832.1 | XP_016882321.1 | 11, 12 | 953 | |||
GenBank mRNA | AB209733.1 | - | 10, 11 | 1021 | ||
AF061832.1 | - | 13, 14 | 1233 | |||
BC000138.2 | - | 12, 13 | 1161 | 2900532 | ||
BC008895.2 | - | 3, 4 | 206 | |||
BC019580.1 | - | 12, 13 | 1129 | 4938259 | ||
L03532.1 | - | 12, 13 | 1128 | |||
L32611.1 | - | 13, 14 | 1012 | |||
X72018.1 | - | 8, 9 | 766 |