Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRICKLE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307979.1 | NP_001294908.1 | 9 | 1408 | ||
NM_006150.4 | NP_006141.2 | 9 | 1485 | |||
GenBank mRNA | AJ011654.1 | - | 9 | 1408 | ||
AK299407.1 | - | 7 | 994 | |||
AK300866.1 | - | 8 | 1310 | |||
AK303200.1 | - | 9 | 1391 | |||
AK303308.1 | - | 8 | 1369 | |||
AK314788.1 | - | 9 | 1436 | |||
BC002468.2 | - | 9 | 1438 | 3347651 | ||
BC016856.1 | - | 9 | 1454 | 3846372 | ||
BT007423.1 | - | 9 | 1382 | |||
CR457211.1 | - | 9 | 1382 |