Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042422.2 | NP_001035887.1 | 2 | 299 | ||
NM_001042423.2 | NP_001035888.1 | 2 | 303 | |||
NM_001206950.1 | NP_001193879.1 | 2 | 315 | |||
NM_001206951.1 | NP_001193880.1 | 2 | 277 | |||
NM_001206952.1 | NP_001193881.1 | 2 | 244 | |||
NM_004207.3 | NP_004198.1 | 2 | 271 | |||
XM_011523607.1 | XP_011521909.1 | 2 | 2192 | |||
GenBank mRNA | AK127319.1 | - | 1 | 2459 | ||
AK223040.1 | - | 2 | 229 | |||
BC112267.1 | - | 1 | 190 | 8327735 | ||
BC112269.1 | - | 1 | 190 | 8327737 | ||
BM926587.1 | - | 2 | 312 | |||
CX872345.1 | - | 2 | 303 | |||
DA373476.1 | - | 2 | 278 | |||
DA419716.1 | - | 2 | 276 | |||
DA583988.1 | - | 2 | 300 | |||
DA895850.1 | - | 2 | 244 | |||
U81800.1 | - | 2 | 211 |