Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146184.1 | NP_001139656.1 | 3 | 533 | ||
NM_001146186.1 | NP_001139658.1 | 1 | 566 | |||
NM_006210.2 | NP_006201.1 | 4 | 609 | |||
GenBank mRNA | AF208967.1 | - | 4 | 579 | ||
AF208968.1 | - | 3 | 503 | |||
AF208970.1 | - | 1 | 567 | |||
BC052616.1 | - | 1 | 208 |
Gene Symbol: ZIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146327.1 | NP_001139799.1 | 4 | 380 | ||
GenBank mRNA | BC104878.1 | - | 3 | 208 | 8143881 |