Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3CB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256045.1 | NP_001242974.1 | 2 | 521 | ||
NM_006219.2 | NP_006210.1 | 11 | 1655 | |||
XM_005247530.2 | XP_005247587.1 | 13 | 2004 | |||
XM_006713659.3 | XP_006713722.1 | 12 | 1875 | |||
XM_011512895.2 | XP_011511197.1 | 14 | 2000 | |||
XM_011512896.1 | XP_011511198.1 | 9 | 1269 | |||
XM_017006619.1 | XP_016862108.1 | 12 | 1678 | |||
XM_017006620.1 | XP_016862109.1 | 11 | 1425 | |||
XM_017006621.1 | XP_016862110.1 | 10 | 1374 | |||
GenBank mRNA | AK293754.1 | - | 2 | 521 | ||
BC114432.1 | - | 11 | 1643 | 40008544 | ||
CR749357.1 | - | 4 | 225 | |||
S67334.1 | - | 11 | 1639 |