Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012328.2 | NP_036460.1 | 3 | 2094 | ||
GenBank mRNA | AB026908.1 | - | 3 | 1927 | ||
AK092204.1 | - | 4 | 1952 |