Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000631.4 | NP_000622.2 | 2, 3 | 302 | ||
NM_013416.3 | NP_038202.2 | 2, 3 | 302 | |||
XM_017028808.1 | XP_016884297.1 | 2, 3 | 666 | |||
GenBank mRNA | AB025219.1 | - | 2, 3 | 154 | ||
AB025220.1 | - | 2, 3 | 154 | |||
AK223324.1 | - | 2, 3 | 273 | |||
AK290924.1 | - | 2, 3 | 224 | |||
BC002798.1 | - | 2, 3 | 302 | 3634177 | ||
BT007346.1 | - | 2, 3 | 118 | |||
BX355636.2 | - | 2, 3 | 278 | |||
CR456528.1 | - | 2, 3 | 199 | |||
CR542078.1 | - | 2, 3 | 118 | |||
X77094.1 | - | 2, 3 | 248 |