Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256106.2 | NP_001243035.1 | 2 | 280 | ||
NM_001256109.2 | NP_001243038.1 | 2 | 280 | |||
NM_001256111.2 | NP_001243040.1 | 2 | 280 | |||
NM_004258.5 | NP_004249.2 | 2 | 280 | |||
XM_017002846.1 | XP_016858335.1 | 3 | 419 | |||
XM_017002847.1 | XP_016858336.1 | 3 | 419 | |||
GenBank mRNA | AK298237.1 | - | 2 | 255 | ||
AK307542.1 | - | 2 | 280 | |||
BC130327.1 | - | 2 | 241 | 40146358 | ||
BC136484.1 | - | 2 | 241 | 9020473 | ||
BC171768.1 | - | 2 | 241 | 9054422 | ||
DA582363.1 | - | 2 | 280 | |||
DA955029.1 | - | 2 | 270 | |||
Z33642.1 | - | 2 | 233 |