Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GP1BA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000173.6 | NP_000164.5 | 2 | 975 | ||
GenBank mRNA | BC027955.1 | - | 2 | 953 | 5210338 | |
J02940.1 | - | 2 | 920 | |||
KC120774.1 | - | 2 | 953 | |||
KC120775.1 | - | 2 | 953 | |||
KC120776.1 | - | 2 | 952 |