Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GRM8 SIRNA Assays ›
Gene Symbol:
GRM8
Gene Name:
glutamate metabotropic receptor 8
Gene Aliases:
GLUR8, GPRC1H, MGLUR8, mGlu8
Chromosome Location:
Chr.7: 126438598 - 127253294 on Build GRCh38
UniGene ID:
Hs.449625
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
144631
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GRM8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000845.2
NP_000836.2
4, 5
1322
NM_001127323.1
NP_001120795.1
5, 6
1568
NR_028041.1
4, 5
1322
XM_006715938.3
XP_006716001.1
5, 6
1642
XM_011516091.1
XP_011514393.1
5, 6
1820
XM_011516092.2
XP_011514394.1
5, 6
1820
XM_011516094.1
XP_011514396.1
5, 6
566
XM_011516095.2
XP_011514397.1
5, 6
699
XM_011516101.2
XP_011514403.1
4, 5
571
XM_011516102.2
XP_011514404.1
5, 6
596
XM_011516103.2
XP_011514405.1
5, 6
728
XM_017012074.1
XP_016867563.1
5, 6
1421
XM_017012075.1
XP_016867564.1
5, 6
772
XM_017012076.1
XP_016867565.1
5, 6
790
XM_017012077.1
XP_016867566.1
4, 5
644
XM_017012078.1
XP_016867567.1
5, 6
669
XM_017012079.1
XP_016867568.1
4, 5
592
XM_017012080.1
XP_016867569.1
3, 4
1248
GenBank mRNA
AJ236921.1
-
4, 5
1026
AJ236922.1
-
4, 5
1026
AK123053.1
-
5, 6
728
AK290197.1
-
5, 6
1619
AK315203.1
-
5, 6
1568
AY608335.1
-
4, 5
1011
BC093725.1
-
4, 5
1037
7939570
BC101675.1
-
4, 5
1037
8069181
BC143344.1
-
4, 5
1037
EU432125.1
-
4, 5
1011
U92459.1
-
4, 5
1068
U95025.1
-
4, 5
1011
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
adenylate cyclase-inhibiting G-protein coupled glutamate receptor signaling pathway
visual perception
sensory perception of smell
negative regulation of cAMP biosynthetic process
regulation of synaptic transmission, glutamatergic
group III metabotropic glutamate receptor activity
G-protein coupled receptor activity
glutamate receptor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0