Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286205.1 | NP_001273134.1 | 11 | 3219 | ||
NM_001286208.1 | NP_001273137.1 | 10 | 3164 | |||
NM_001286209.1 | NP_001273138.1 | 11 | 3111 | |||
NM_001481.2 | NP_001472.1 | 11 | 3096 | |||
XM_005256304.4 | XP_005256361.1 | 12 | 3751 | |||
XM_006721175.3 | XP_006721238.1 | 11 | 3149 | |||
XM_011522990.2 | XP_011521292.1 | 11 | 3170 | |||
XM_011522991.1 | XP_011521293.1 | 12 | 3325 | |||
XM_011522992.2 | XP_011521294.1 | 10 | 3118 | |||
XM_017023122.1 | XP_016878611.1 | 13 | 3476 | |||
XM_017023123.1 | XP_016878612.1 | 11 | 3175 | |||
XM_017023124.1 | XP_016878613.1 | 10 | 2962 | |||
XM_017023125.1 | XP_016878614.1 | 10 | 3094 | |||
GenBank mRNA | AF050079.1 | - | 11 | 3096 | ||
BX647947.1 | - | 10 | 3083 | |||
CR749499.1 | - | 9 | 3775 |