Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FPR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002030.4 | NP_002021.3 | 2 | 2206 | ||
XM_011526687.2 | XP_011524989.1 | 2 | 2097 | |||
GenBank mRNA | AK123030.1 | - | 3 | 2149 | ||
BC059388.1 | - | 2 | 2189 | 30332008 |
Set Membership: |
G-protein coupled receptor |