Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GYPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308187.1 | NP_001295116.1 | 6 | 1729 | ||
NM_001308190.1 | NP_001295119.1 | 6 | 1669 | |||
NM_002099.7 | NP_002090.4 | 7 | 1768 | |||
XM_017008134.1 | XP_016863623.1 | 6 | 1693 | |||
XM_017008135.1 | XP_016863624.1 | 8 | 1835 | |||
XM_017008136.1 | XP_016863625.1 | 5 | 1658 | |||
GenBank mRNA | BC013328.1 | - | 7 | 1643 | 4134368 | |
BX649043.1 | - | 1 | 1640 | |||
L31860.1 | - | 7 | 1661 | |||
X51798.1 | - | 7 | 1634 |