Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002155.4 | NP_002146.2 | 1 | 1828 | ||
GenBank mRNA | AK093925.1 | - | 1 | 1828 | ||
AK223362.1 | - | 1 | 1828 | |||
AK295212.1 | - | 2 | 1751 | |||
AK312756.1 | - | 1 | 1828 | |||
BC004279.1 | - | 1 | 215 | |||
BC035665.1 | - | 1 | 1818 | 5723718 | ||
X51758.1 | - | 1 | 647 |