Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002156.4 | NP_002147.2 | 2 | 280 | ||
NM_199440.1 | NP_955472.1 | 2 | 260 | |||
GenBank mRNA | AK298368.1 | - | 2 | 199 | ||
AK298632.1 | - | 2 | 230 | |||
AK298844.1 | - | 2 | 199 | |||
AK301276.1 | - | 2 | 235 | |||
AK312240.1 | - | 2 | 213 | |||
AU098504.1 | - | 2 | 282 | |||
BC002676.2 | - | 2 | 190 | 3609039 | ||
BC003030.1 | - | 2 | 189 | 2821157 | ||
BC047350.1 | - | 1 | 350 | |||
BC067082.1 | - | 2 | 188 | 4025725 | ||
BC073746.1 | - | 2 | 188 | 6372068 | ||
BG701476.1 | - | 2 | 281 | |||
BX647145.1 | - | 2 | 268 | |||
EU719626.1 | - | 1 | 143 | |||
KU178139.1 | - | 1 | 143 | |||
KU178141.1 | - | 1 | 143 | |||
M22382.1 | - | 2 | 188 | |||
M34664.1 | - | 2 | 167 |