Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002187.2 | NP_002178.2 | 8 | 1647 | ||
GenBank mRNA | AY046592.1 | - | 2 | 675 | ||
AY046593.1 | - | 2 | 675 | |||
M65290.1 | - | 7 | 1618 |