Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297649.1 | NP_001284578.1 | 5 | 486 | ||
NM_004356.3 | NP_004347.1 | 5 | 604 | |||
XM_011520492.2 | XP_011518794.1 | 5 | 838 | |||
XM_017018598.1 | XP_016874087.1 | 5 | 347 | |||
GenBank mRNA | AB209380.1 | - | 2 | 3271 | ||
BC002978.1 | - | 5 | 411 | 3510634 | ||
BC093047.1 | - | 5 | 602 | 30398483 | ||
BG397506.1 | - | 6 | 474 | |||
BP323362.1 | - | 5 | 500 | |||
BQ709484.1 | - | 601 | ||||
BT019507.1 | - | 5 | 371 | |||
BT019508.1 | - | 5 | 371 | |||
M33680.1 | - | 5 | 609 |