Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004119.2 | NP_004110.2 | 8 | 1059 | ||
NR_130706.1 | 8 | 1059 | ||||
XM_011535015.2 | XP_011533317.1 | 8 | 1058 | |||
XM_011535017.2 | XP_011533319.1 | 8 | 961 | |||
XM_011535018.2 | XP_011533320.1 | 5 | 569 | |||
XM_017020486.1 | XP_016875975.1 | 7 | 838 | |||
XM_017020487.1 | XP_016875976.1 | 9 | 1132 | |||
XM_017020488.1 | XP_016875977.1 | 7 | 825 | |||
XM_017020489.1 | XP_016875978.1 | 7 | 920 | |||
GenBank mRNA | BC036028.1 | - | 8 | 1059 | ||
BC126350.1 | - | 8 | 1043 | 8992066 | ||
BC144039.1 | - | 8 | 1044 | 9052559 | ||
BC144040.1 | - | 8 | 1044 | 9052560 | ||
U02687.1 | - | 8 | 1034 | |||
Z26652.1 | - | 8 | 977 |