Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256486.1 | NP_001243415.1 | 8 | 937 | ||
NM_001256487.1 | NP_001243416.1 | 8 | 965 | |||
NM_001256488.1 | NP_001243417.1 | 7 | 842 | |||
NM_004487.4 | NP_004478.3 | 8 | 922 | |||
XM_005247371.4 | XP_005247428.1 | 8 | 1397 | |||
XM_005247372.4 | XP_005247429.1 | 8 | 973 | |||
XM_005247373.1 | XP_005247430.1 | 7 | 789 | |||
XM_006713587.1 | XP_006713650.1 | 8 | 912 | |||
XM_006713588.1 | XP_006713651.1 | 8 | 897 | |||
XM_006713589.1 | XP_006713652.1 | 8 | 894 | |||
XM_006713590.1 | XP_006713653.1 | 7 | 789 | |||
XM_006713591.1 | XP_006713654.1 | 6 | 672 | |||
XM_011512699.2 | XP_011511001.1 | 8 | 864 | |||
XM_017006189.1 | XP_016861678.1 | 8 | 1382 | |||
XM_017006190.1 | XP_016861679.1 | 8 | 1549 | |||
XM_017006191.1 | XP_016861680.1 | 7 | 1274 | |||
XM_017006192.1 | XP_016861681.1 | 8 | 958 | |||
XM_017006193.1 | XP_016861682.1 | 8 | 955 | |||
XM_017006194.1 | XP_016861683.1 | 7 | 850 | |||
XM_017006195.1 | XP_016861684.1 | 2 | 819 | |||
GenBank mRNA | AB371588.1 | - | 8 | 910 | ||
AB593126.1 | - | 7 | 816 | |||
AK296903.1 | - | 4 | 396 | |||
AK298986.1 | - | 5 | 488 | |||
AK300437.1 | - | 8 | 911 | |||
BC065301.1 | - | 7 | 713 | |||
D25542.1 | - | 8 | 920 | |||
X75304.1 | - | 8 | 910 |