Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288824.2 | NP_001275753.1 | 3 | 725 | ||
NM_001288825.2 | NP_001275754.1 | 4 | 841 | |||
NM_001288826.2 | NP_001275755.1 | 4 | 701 | |||
NM_001288827.2 | NP_001275756.1 | 4 | 698 | |||
NM_001288828.2 | NP_001275757.1 | 4 | 621 | |||
NM_005260.4 | NP_005251.1 | 2 | 672 | |||
XM_005271957.4 | XP_005272014.1 | 3 | 1662 | |||
XM_006714585.3 | XP_006714648.1 | 3 | 1595 | |||
XM_011543308.2 | XP_011541610.1 | 3 | 2643 | |||
XM_011543309.1 | XP_011541611.1 | 4 | 605 | |||
XM_011543310.1 | XP_011541612.1 | 4 | 553 | |||
XM_011543311.2 | XP_011541613.1 | 2 | 1418 | |||
GenBank mRNA | AK301962.1 | - | 4 | 668 | ||
AK315973.1 | - | 3 | 725 | |||
BC096228.3 | - | 2 | 672 | 40005256 | ||
BC096229.3 | - | 2 | 672 | 40005257 | ||
BC096230.3 | - | 2 | 672 | 40005258 | ||
BC096231.1 | - | 2 | 672 | 40005259 |