Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005295.2 | NP_005286.2 | 3 | 1437 | ||
XM_011516056.2 | XP_011514358.1 | 2 | 1123 | |||
GenBank mRNA | AB209270.1 | - | 1 | 347 | ||
AK122621.1 | - | 3 | 1434 | |||
AK307393.1 | - | 3 | 1434 | |||
BC107128.2 | - | 1 | 91 | 40024013 | ||
BX647649.1 | - | 2 | 301 | |||
HQ995527.1 | - | 1 | 91 |
Set Membership: |
G-protein coupled receptor |