Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111112.1 | NP_001104582.1 | 6 | 1130 | ||
NM_001111113.1 | NP_001104583.1 | 5 | 1106 | |||
NM_001312646.1 | NP_001299575.1 | 5 | 1076 | |||
NM_001312647.1 | NP_001299576.1 | 7 | 1171 | |||
NM_001312648.1 | NP_001299577.1 | 6 | 1165 | |||
NM_005339.4 | NP_005330.1 | 7 | 1259 | |||
GenBank mRNA | AK093533.1 | - | 7 | 1080 | ||
AK291454.1 | - | 5 | 1011 | |||
AK295846.1 | - | 5 | 1011 | |||
AK307130.1 | - | 6 | 1070 | |||
U58522.1 | - | 7 | 978 |