Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005546.3 | NP_005537.3 | 9 | 868 | ||
XM_017009443.1 | XP_016864932.1 | 7 | 481 | |||
GenBank mRNA | AB209622.1 | - | 9 | 921 | ||
AK312846.1 | - | 9 | 837 | |||
BC109077.2 | - | 9 | 859 | 40034274 | ||
BC109078.1 | - | 9 | 859 | 40034276 | ||
D13720.1 | - | 9 | 868 |