Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012202.4 | NP_036334.1 | 3 | 676 | ||
XM_006718500.2 | XP_006718563.1 | 4 | 663 | |||
GenBank mRNA | AF075042.1 | - | 2 | 339 | ||
AF087900.1 | - | 3 | 613 | |||
AF092129.1 | - | 3 | 613 | |||
BC015563.2 | - | 3 | 557 | 4124714 |