Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR6A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278546.1 | NP_001265475.1 | 10 | 2073 | ||
NM_001489.4 | NP_001480.3 | 10 | 2070 | |||
NM_033334.3 | NP_201591.2 | 10 | 2085 | |||
XM_005251917.4 | XP_005251974.1 | 10 | 2031 | |||
XM_005251918.4 | XP_005251975.1 | 9 | 3254 |