Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000188.2 | NP_000179.2 | 8 | 1021 | ||
NM_001322364.1 | NP_001309293.1 | 10 | 1114 | |||
NM_001322365.1 | NP_001309294.1 | 13 | 1419 | |||
NM_001322366.1 | NP_001309295.1 | 8 | 912 | |||
NM_001322367.1 | NP_001309296.1 | 7 | 925 | |||
NM_033496.2 | NP_277031.1 | 8 | 1018 | |||
NM_033497.2 | NP_277032.1 | 11 | 1236 | |||
NM_033498.2 | NP_277033.1 | 11 | 1290 | |||
NM_033500.2 | NP_277035.2 | 12 | 1383 | |||
XM_005269737.1 | XP_005269794.1 | 8 | 1292 | |||
XM_011539732.1 | XP_011538034.1 | 12 | 1290 | |||
GenBank mRNA | AB209526.1 | - | 8 | 1017 | ||
AK091267.1 | - | 8 | 1146 | |||
AK128226.1 | - | 13 | 1414 | |||
AK292012.1 | - | 8 | 1021 | |||
AK294428.1 | - | 8 | 1018 | |||
AK309904.1 | - | 7 | 847 | |||
BC008730.2 | - | 8 | 976 | 3163058 | ||
M75126.1 | - | 8 | 1001 | |||
X66957.1 | - | 2 | 62 |